Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MT2-MMP, Catalytic Domain, Human, Recombinant Protein, E. coli

Catalog No. 47593810UG
Click to view available options
Quantity:
10 μg
Recombinant, human pro-MT2-MMP expressed in E. coli as a periplasmic protein and activated to yield the active catalytic domain (amino acids 91-267). The catalytic domain is comprised of amino acid residues Tyr91 to Gly267 of mature human MT2-MMP. MT2-MMP is used to study the activation of MMP-2 and the degradation of extracellular matrix proteins.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 50mM Tris-HCl, 5mM CaCl2, pH 7.5.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen MT2-MMP
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.