Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ MultiScreenHTS 384-Well Filter Plates
Use in automated, high throughput applications, including biological assays and sample preparation. MultiScreenHTS 384-Well Filter Plates are non-sterile, single use plates available in several mesh materials and pore sizes to meet the needs of different applications.
Supplier: MilliporeSigma™ MZHVN0W10
Description
For use in receptor-ligand binding, general sample preparation, enzyme assays with precipitation and DNA purification.- Rigid sidewalls allows full compatibility with automated/robotic systems
- Unique plate design and individually sealed filter discs prevent cross-talk between wells
- Opaque plastic lids prevents light transfer between wells during direct scintillation counting
- Barcoding available
- Meet ANSI™/SBS standards
Specifications
Standard | |
16 x 24 | |
Opaque, White Bottom | |
384 | |
0.09cm2 | |
10/Pk. | |
1.28 cm | |
1.44 cm |
Durapore PVDF | |
0.45 μm | |
Styrene | |
Non-sterile | |
Receptor binding assays on whole cells and cell fragments, protein kinase, phosphatase precipitation assays, bead, resin based assays | |
1.28 x 8.55 x 1.44 cm | |
8.55 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction