Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane

Catalog No. MSHAN4510
Click to view available options
Color:
Clear
Opaque
Material:
Barex
Styrene
Sterility:
Non-sterile
Sterile
Quantity:
10/Pk.
50/Pk.

Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies

  • Developed for high-throughput use with automated work stations
  • Individually sealed wells prevent incubation crosstalk
  • Removable underdrain allows access to the filters
  • Rigid sidewalls improve handling
  • Ample surfaces provided for bar code labels
  • Pore size: .45μm

Solid-phase RIA, DNA hybridization, ELISA and protein binding

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Mixed Cellulose Esters
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Product Type MultiScreenHTS-HA Filter Plate
Material Styrene
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28 cm2
For Use With (Application) Solid phase RIA, DNA hybridization, ELISA, protein binding
Quantity 10/Pk.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.