Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane

Catalog No. p-3740868
Click to view available options
Product Type:
Microplate 8-Well Strip
Microplate Strip Support Frame

Includes

Twelve 8-well strips in a support frame. Additional support frames also available separately.

8-well strips enable processing of less than 96 samples at one time

  • For ELISpot and protein binding assays

TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Array 1 x 8
Color Clear
Product Type Microplate 8-Well Strip
Material ABS Plastic
No. of Wells 8
Sterility Sterile
For Use With (Application) ELIspot and protein binding assays
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.