Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates

Catalog No. MSFCN6B10 Shop All MilliporeSigma Products
Click to view available options
Pore Size:
1.2/0.65 μm

Catalog No. MSFCN6B10

Supplier: MilliporeSigma™ MSFCN6B10

Only null left
Add to Cart
Add to Cart

Receptor/ligand binding assays, sample prep,TCA precipitation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Format Standard
Membrane Hydrophilic PVDF
Array 8 x 12
Pore Size 1.2/0.65 μm
Color Opaque
Material Barex
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28cm2
For Use With (Application) Receptor, ligand binding assays, sample prep, enzyme assays with precipitation, DNA purification
Quantity 10/Pk.
Dimensions (L x W x H) 0.5 x 8.55 x 0.56 in. (1.28 x 21.71 x 1.44 cm)
Length (Metric) 1.28 cm
Width (Metric) 21.71 cm
Height (Metric) 1.44 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.