Learn More
MilliporeSigma™ MultiScreen™HTS Vacuum Manifold
For vacuum filtration using 96- and 384-well MultiScreenHTS, MultiScreen™ Solvinert, and Solvinert Deep Well filter plates
Supplier: MilliporeSigma™ MSVMHTS00
Includes: Manifold base, standard collar, gaskets and gasket inserts, tubing, valves, pressure gauge
Description
- Accepts deep well or standard receiver plates Suitable for use with manual or automated protocols; ANSI/SBS compliant footprint allows for easy robotic deck integration
- Easily configured for both flow-to-waste and analyte collection
- DirectStack™ technology for crosstalk-free filtrate collection
- Manifold components are resistant to solvents
Specifications
MultiScreen Vacuum Manifold |