Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NADPH-P450 Oxidoreductase, Human, Recombinant Protein, S. frugiperda

Supplier:  MilliporeSigma™ 481974200UG

Catalog No. 48-197-4200UG

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Recombinant, human NADPH-P450 oxidoreductase expressed in S. frugiperda insect cells. A membrane bound flavoprotein containing one mole each of FAD and FMN. Transfers electrons from NADPH to the various isoforms of cytochrome P450. Typically used at ratio 1:10 to 1:2 (reductase:P450) for in vitro reconstitution assays. Suitable for in vitro drug metabolism and toxicology research including reconstituted P450-dependent biotransformation reactions.
Specifications

Specifications

Concentration on Label
Aqueous buffers
200μg
NADPH-P450 Oxidoreductase
RUO
Recombinant
≥90%
In 10mM potassium phosphate buffer, 1mM DTT, 100μM EDTA, 20% glycerol, pH 7.7.
SDS-PAGE
S. frugiperda
Following initial thaw, aliquot and freeze (−70°C).
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit