Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Natriuretic Peptide, C-Type, Human and Porcine, Calbiochem
$241.00 - $412.00
Specifications
Species | Human, Porcine |
---|---|
Purification Method | HPLC |
Purity | ≥96% |
CAS | 127869-51-6 |
Color | White to Off-white |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
05-230-31005M
|
MilliporeSigma™
052303100.5MG |
0.5 mg |
Each for $241.00
|
|
|||||
05-230-3101MG
|
MilliporeSigma™
052303101MG |
1 mg |
Each for $412.00
|
|
|||||
Description
Member of the natriuretic peptide family that shares structural homology with ANP and plays a role in the regulation of body fluid homeostasis, vascular tone, and vascular growth. Inhibits adenylate cyclase activity in a concentration-dependent manner. Stimulates particulate guanylate cyclase activity. Shown to exert neuromodulatory effects independently of guanylate cyclase activation or adenylate cyclase inhibition.Specifications
Human, Porcine | |
≥96% | |
White to Off-white | |
Lyophilized Solid | |
Natriuretic Peptide, C-Type |
HPLC | |
127869-51-6 | |
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 1 month at −20°C. | |
Sold on the basis of peptide content. | |
RUO - research use only |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title