Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Neuropeptide Y, Human, Calbiochem
Supplier: MilliporeSigma™ 052320050.5MG
Description
A potent vasoconstrictor. Reversibly inhibits Ca2+-activated K+ channels in vascular smooth muscle cells.Specifications
Human | |
≥97% | |
White to Off-white | |
Lyophilized Solid | |
Neuropeptide Y | |
RUO - research use only |
HPLC | |
90880-35-6 | |
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. | |
Supplied as trifluoroacetate salt. Sold on the basis of peptide content. | |
0.5 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title