Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Neuropeptide Y, Human, Calbiochem

$427.00 - $641.00

Specifications

Species Human
Purification Method HPLC
Purity ≥97%
CAS 90880-35-6
Color White to Off-white
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05-232-0051MG
View Documents
MilliporeSigma™
052320051MG
1 mg
Each for $641.00
Add to Cart
 
05-232-00505M
View Documents
MilliporeSigma™
052320050.5MG
0.5 mg
Each for $427.00
Add to Cart
 
Description

Description

A potent vasoconstrictor. Reversibly inhibits Ca2+-activated K+ channels in vascular smooth muscle cells.
Specifications

Specifications

Human
≥97%
White to Off-white
Lyophilized Solid
Neuropeptide Y
HPLC
90880-35-6
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C.
Supplied as trifluoroacetate salt. Sold on the basis of peptide content.
RUO - research use only
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit