Learn More
EMD Millipore Novagen™ NovaBlue GigaSingles™ Competent Cells Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Millipore Sigma 712274
 View more versions of this product
Catalog No. 71-227-4
  Check Availability
Add to cart
Specifications
Description
NovaBlue GigaSingles™ Competent Cells produce greater than 1 x 109 colonies/ug plasmid DNA for cloning applications requiring high-efficiency transformations.
Description & Specifications
Specifications
Guaranteed efficiency: >1 x 109 cfu/μg
22 reactions
Store at ≤-70deg.C