Learn More
EMD Millipore Novagen™ NovaBlue Singles™ Competent Cells Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Millipore Sigma 701814
 View more versions of this product
Catalog No. 70-181-4
  Check Availability
Add to cart
Specifications
Description
NovaBlue Singles Competent Cells, like all Novagen Singles Competent Cells, are designed for ultimate convenience and reliability in plasmid transformation. The cells are grown and made chemically competent by an optimized procedure. Cells are provided in 50µL volumes that eliminate the need to aliquot, freeze/thaw, or waste partially used vials, thus saving time and increasing performance. For use, simply thaw, add DNA, incubate five minutes on ice, heat shock for 30 seconds, place on ice for two minutes, add SOC Medium, and plate directly (when selecting for ampicillin resistance) or after incubation at 37°C for 30 minutes (when selecting for kanamycin resistance).
Description & Specifications
Specifications
Guaranteed efficiency: >1.5 x 108 cfu/μg
22 reactions
Store at ≤-70deg.C