Learn More
EMD Millipore Novagen™ Rosetta™ Competent Cells Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Millipore Sigma 709534
 View more versions of this product
Catalog No. 70-953-4
  Check Availability
Add to cart
Specifications
Description
Rosetta™ host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli. These strains supply tRNAs for AGG, AGA, AUA, CUA, CCC, GGA codons on a compatible chloramphenicol-resistant plasmid. Thus the Rosetta strains provide for universal translation which is otherwise limited by the codon usage of E. coli. The tRNA genes are driven by their native promoters.
Description & Specifications
Specifications
Guaranteed efficiency: >2 x 106 cfu/μg
1mL
Store at ≤-70deg.C