Learn More
MilliporeSigma™ NRG1-β1/HRG1-β1 ECD, Human, Recombinant Protein
Manufacturer:  MilliporeSigma™ 49202850UG
Catalog No. 49-202-850UG
Add to cart
Description
Description
Recombinant, human neuregulin 1-β1/heregulin 1-β1 extracellular domain (NRG1-beta1/HRG1-beta1 ECD) consisting of amino acids 2-246 and expressed in E. coli. All NRG1 isoforms contain an EGF-like domain (β-splice variant) that is used to directly bind to the ErbB3 or ErbB4 receptor tyrosine kinases.
Specifications
Specifications
Lyophilized
Reconstitute 50 to 100μg/mL with sterile PBS containing ≥1% human or bovine serum albumin.
≥90%
NRG1-β1/HRG1-β1 ECD
RUO
Yes
Lyophilized from 0.2μM filtered acetonitrile, TFA, BSA.
SDS-PAGE
50μg
Following reconstitution refrigerate (4°C) for short term-storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Human