Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Steriflip™ Sterile Centrifuge Tube Top Filter Units

Sterile, disposable filter unit for cell isolation.

$307.00

Specifications

Color Clear
Connectors Double lead thread with vacuum port
Disposable Single-use
Material MBS/Polypropylene
Sterility Sterile
View More Specs

Products 4
Catalog Number Mfr. No. Material (Membrane) Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Material (Membrane) Pore Size Price Quantity & Availability  
SCNY00020
View Documents Promotion Details
MilliporeSigma™
SCNY00020
Nylon Mesh 20 μm
Pack of 25 for $307.00
Add to Cart
 
SCNY00040
View Documents Promotion Details
MilliporeSigma™
SCNY00040
Nylon Net 40 μm
Pack of 25 for $307.00
Edge
Add to Cart
 
SCNY00060
View Documents Promotion Details
MilliporeSigma™
SCNY00060
Nylon Mesh 60 μm
Pack of 25 for $307.00
Add to Cart
 
SCNY00100
View Documents Promotion Details
MilliporeSigma™
SCNY00100
Nylon Net 100 μm
Pack of 25 for $307.00
Edge
Add to Cart
 
Description

Description

Overview
The Steriflip filter unit is designed to work with standard 50 mL centrifuge tubes, which eliminates sample transfer. To filter, just attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply the vacuum. When finished, the filtrate collects in the attached 50 mL centrifuge tube.

New pore sizes ideal for Cell Isolation applications

Steriflip Nylon Net filters are now available in 40, 60 and 100 μm pore sizes, making them ideal for stem cell, cardiomyocite, neurological and skeletal tissue applications. The vacuum-assisted, closed system option ensures sample sterility while enabling faster separation of large volumes of cellular material and improved recoveries.

The Steriflip™ filter unit eliminates messy pouring during sample transfer. Directly attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply vacuum. The filtrate collects in the attached 50 mL centrifuge tube. Steriflip™ filters come in different pore sizes that are ideal for cell isolation. These include nylon net filters in 40, 60 and 100 μm pore sizes, which are ideal for stem cell, cardiomyocyte, neurological and skeletal tissue applications. The vacuum-assisted, closed system ensures sample sterility while enabling fast separation of large volumes of cellular material for efficient recoveries.

Features & Benefits

  • Eliminates messy sample transfer via a simple vacuum-assisted, closed system
  • Isolate cells from cellular debris using any of the various pore-sized nylon net filters
  • Ideal for sterilizing small batches of culture media
  • Attaches to any standard 50 mL centrifuge tube, eliminating unnecesary materials, time and risk of spillage

Applications: Sterilization of Aqueous Solutions (up to 50 mL), Sterile Filtration of Small Batches of Culture Media, Sterile Filtration of Buffers with High Value Protein or Small Molecule Components, Cell Isolation

Specifications

Specifications

Clear
Single-use
Sterile
Removal of coarse particulates, laboratory filtration
25 Pack
50 mL
13.8 cm
Double lead thread with vacuum port
MBS/Polypropylene
7 cm2
25Pack
Steriflip-NY Filter Unit
5.43 in.
6.4 cm
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit