Learn More
MilliporeSigma™ Omnipore™ Membrane Filter Available on GSA/VA Contract for Federal Government customers only.
Compatible with virtually all solvents, acids and alkaline solutions
$303.00 - $401.00
Specifications
Color White
Material Polytetrafluoroethylene (PTFE)
Quantity (No. per Case) 100/pk.
View More Specs
Products 6
Catalog Number Mfr. No. Diameter (Metric) Outer Pore Size Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Outer Pore Size Price Quantity & Availability  
JGWP04700 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
JGWP04700
47mm 0.2μm
Pack of 100 for $401.00
Add to cart
JGWP09025
View Documents
MilliporeSigma™
JGWP09025
90mm 0.2μm
Each for $333.00
Add to cart
JGWP02500
View Documents
MilliporeSigma™
JGWP02500
25mm 0.2μm
Pack for $303.00
Add to cart
JHWP02500 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
JHWP02500
25mm 0.45μm
Pack for $303.00
Add to cart
JHWP09025
View Documents
MilliporeSigma™
JHWP09025
90mm 0.45μm
Each for $333.00
Add to cart
JHWP04700 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
JHWP04700
47mm 0.45μm
Pack for $401.00
Add to cart
Description
Description
  • White, plain (non-gridded) filter surface
  • 65µm thick, 80% porosity
  • Hydrophilic PTFE membrane bonded to a high-density polyethylene support
  • Maximum operating temperature of 130°C
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
White
Polytetrafluoroethylene (PTFE)
100/pk.