Learn More
Description
- High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
- On-board PES membrane prefilter for extended throughput in high fouling media
- Robust construction
- Withstand multiple sterilization cycles
- Easy wetting and integrity testing
- Broad chemical compatibility across a wide pH range (pH 1–14)
- Hydrophilic Polyethersulfone filter
- Polyester supports
- Structural components: Gamma stable polypropylene/Polysulfone
- Silicone vent O-rings
- Gamma compatible sterilization
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Specifications
Specifications
| Filter Type | Membrane Filter |
| Material (Membrane) | PES |
| Pore Size | 0.5/0.2 μm |
| Sterility | Non-sterile |
| Filtration Area | 0.014 m2 |
| For Use With (Application) | Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals |
| Housing Material | Polypropylene |
| Inlet Style | 0.75 in. (19mm) sanitary flange |
| Outlet Style | 0.75 in. (19mm) sanitary flange |
| Type | Filter Capsule |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.