Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Opticap™ Sterile XL150 Capsule Filters

For sterilizing grade performance and mycoplasma removal

$437.00 - $654.00

Specifications

For Use With (Application) Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Length (English) 3.8 in.
Length (Metric) 9.7 cm
View More Specs

Products 6
Catalog Number Mfr. No. Pore Size Filtration Area Inlet Style Outlet Style Price Quantity & Availability  
Catalog Number Mfr. No. Pore Size Filtration Area Inlet Style Outlet Style Price Quantity & Availability  
KHGES015HH3
View Documents
MilliporeSigma™
KHGES015HH3
0.5/0.2 μm 0.014 m2 0.56 in. (14mm) hose barb 0.56 in. (14mm) hose barb
Pack of 3 for $449.00
Add to Cart
 
KHGES015FF3
View Documents
MilliporeSigma™
KHGES015FF3
0.5/0.2 μm 0.014 m2 0.75 in. (19mm) sanitary flange 0.56 in. (14mm) hose barb
Pack of 3 for $437.00
Add to Cart
 
KHGES015FH3
View Documents
MilliporeSigma™
KHGES015FH3
0.5/0.2 μm 0.014 m2 0.75 in. (19mm) sanitary flange 0.75 in. (19mm) sanitary flange
Pack of 3 for $450.00
Add to Cart
 
KHVES015HH3
View Documents
MilliporeSigma™
KHVES015HH3
0.5/0.01 μm 0.02 m2 0.56 in. (14mm) hose barb 0.56 in. (14mm) hose barb
Pack of 3 for $654.00
Add to Cart
 
KHVES015FH3
View Documents
MilliporeSigma™
KHVES015FH3
0.5/0.01 μm 0.02 m2 0.75 in. (19mm) sanitary flange 0.56 in. (14mm) hose barb
Pack of 3 for $555.50
Add to Cart
 
KHVES015FF3
View Documents
MilliporeSigma™
KHVES015FF3
0.5/0.01 μm 0.02 m2 0.75 in. (19mm) sanitary flange 0.75 in. (19mm) sanitary flange Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
Description

Description

  • High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
  • On-board PES membrane prefilter for extended throughput in high fouling media
  • Robust construction
  • Withstand multiple sterilization cycles
  • Easy wetting and integrity testing
  • Broad chemical compatibility across a wide pH range (pH 1–14)
  • Hydrophilic Polyethersulfone filter
  • Polyester supports

  • Structural components: Gamma stable polypropylene/Polysulfone
  • Silicone vent O-rings
  • Gamma compatible sterilization

Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals

Specifications

Specifications

Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
3.8 in.
9.7 cm
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.