Learn More
MilliporeSigma™ Opticap™ Sterilizing Grade XL2 Durapore Capsule Filters
ideal for sterile filtration of cell culture media, protein solutions and other high fouling fluids
Supplier:  MilliporeSigma™ KVGLA02FF3
 View more versions of this product
Catalog No. KVGLA02FF3
Add to Cart
Description
Description
Specifications
Specifications
Inlet/Outlet Connection: 19mm Sanitary Flange
0.22 μm
Product Suggestions
Product Suggestions