Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ OptiScale™ Capsule Filters for Aseptic Processing Applications

Small-volume option for process screening and scaling

Manufacturer:  MilliporeSigma™ SVGLA47HH3

 View more versions of this product

Catalog No. SVGLA47HH3


Add to cart

Description

Description

  • With “drop-in” filters
  • Non-sterile
  • PVDF vent cap
  • Wettability: Hydrophilic
  • Housing and drainage disk: Polycarbonate
  • Filter support: Polypropylene
  • Filtration Area: 17.7cm2

Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates

Specifications

Specifications

Non-sterile
Polycarbonate
3.7 in.
17.7cm2
0.22μm
Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates
9.4cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit