Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ OptiScale™ Capsule Filters for Aseptic Processing Applications

Catalog No. p-3742998
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.22 μm
0.45/0.22 μm
0.5/0.2 μm
Length (English):
2.9 in.
2.91 in.
3.2 in.
3.24 in.
3.7 in.
Length (Metric):
7.4 cm
8.2 cm
9.4 cm

Small-volume option for process screening and scaling

  • With “drop-in” filters
  • Non-sterile
  • PVDF vent cap
  • Wettability: Hydrophilic
  • Housing and drainage disk: Polycarbonate
  • Filter support: Polypropylene
  • Filtration Area: 17.7cm2

Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates

TRUSTED_SUSTAINABILITY

Specifications

Pore Size 0.5/0.2 μm
Sterility Non-sterile
Filtration Area 17.7 cm2
For Use With (Application) Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates
Housing Material Polycarbonate
Length (English) 3.7 in.
Length (Metric) 9.4 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.