Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ PACAP 27 Amide, Ovine neuropeptide, Calbiochem
Supplier: MilliporeSigma™ 052321510.5MG
Description
Increases cAMP levels in a dose-dependent manner (EC50 = 4.7nM). Increases tyrosine hydroxylase expression in chromaffin cells.Specifications
Ovine | |
≥98% | |
White to Off-white | |
Solid | |
PACAP 27 Amide | |
RUO - research use only |
HPLC | |
127317-03-7 | |
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 6 months at −20°C. | |
Supplied as a trifluoroacetate salt. | |
0.5 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title