Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ PACAP 38, Ovine neuropeptide, Calbiochem
$170.00 - $751.00
Specifications
Species | Ovine |
---|---|
Purification Method | HPLC |
Purity | ≥96% |
CAS | 137061-48-4 |
Color | White to Off-white |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
05-232-15001M
|
MilliporeSigma™
052321500.1MG |
0.1 mg |
Each for $170.00
|
|
|||||
05-232-1501MG
|
MilliporeSigma™
052321501MG |
1 mg |
Each for $751.00
|
|
|||||
Description
More active than vasoactive intestinal peptide (VIP) in stimulating adenylate cyclase (EC50 = 7nM).Specifications
Ovine | |
≥96% | |
White to Off-white | |
Solid | |
RUO - research use only |
HPLC | |
137061-48-4 | |
Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 3 months at −20°C. | |
PACAP 38 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title