Learn More
MilliporeSigma™ pET Expression System 11
Manufacturer:  MilliporeSigma™ 694053
Catalog No. 69-405-3
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The pET Expression System 11 contains 10μg each of the four versions of pET-11 (pET-11a-d). The pET-11a-d vectors carry an N-terminal T7·Tag™ sequence and BamH I cloning site. These vectors are the precursors to many pET family vectors; the pET-21a-d(+) series corresponds to pET-11a-d but incorporates several additional features.
Specifications
Specifications
Target gene cloning and expression
BamH I; Nde I cloning site (a, b, and c versions); Nco I site (d version)