Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Phosphodiesterase 10A2, HisTag™, Human, Recombinant Protein, S. frugiperda
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 U
Description
Full-length, recombinant, human, phosphodiesterdase 10A2 (accession no. 10A2) fused at the N-terminus to a His·Tag7reg; sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. Supplied as a partially purified preparation of catalytically active enzyme.
Specifications
Specifications
| Formulation | In RIPA buffer (150mM NaCl, 10mM Tris-HCl, 1% NP-40), pH 8.3. |
| Quantity | 10 U |
| Source | S. frugiperda |
| Immunogen | Phosphodiesterase 10A2 |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Conjugate | His-Tag |
| Species | Human |
| Recombinant | Recombinant |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction