Learn More
MilliporeSigma™ pIEx-2 Ek/LIC Vector Kit
Manufacturer:  MilliporeSigma™ 712403
Catalog No. 71-240-3
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The pIEx™-2 Ek/LIC Vector is designed to allow rapid, high-level protein expression in insect cells without the time-consuming process of creating a recombinant baculovirus. This vector features the hr5 enhancer and the IE1 (immediate early) promoter to direct expression in insect cells. This promoter/enhancer combination recruits endogenous insect cell transcription machinery, thereby avoiding baculovirus infection and the associated cytotoxic effects.
Specifications
Specifications
Rapid, high-level protein expression in insect cells without creating a recombinant baculovirus
IE1
20 Reactions