Learn More
MilliporeSigma™ PlGF-2, Mouse, Recombinant Protein
Manufacturer:  MilliporeSigma™ 50617110UG
Catalog No. 50-617-110UG
Add to cart
Description
Description
Recombinant, mouse placenta growth factor 2 (PlGF-2), consisting of amino acids 24-158 and 27-158, and expressed in S. frugiperda Sf21 cells using a baculovirus expression system. PlGF-2 is a member of the VEGF family of growth factors.
Specifications
Specifications
Lyophilized
Reconstitute 50 to 100μg/mL with sterile PBS contanining ≥1% human or bovine serum albumin.
≥97%
PlGF-2
RUO
Yes
Lyophilized from 0.2μM filtered acetonitrile, TFA, BSA.
SDS-PAGE
10μg
Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Mouse