Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ PP2A1, Bovine Kidney, Protein
Supplier: MilliporeSigma™ 5395085UG
Description
Native, PP2A1 from bovine kidney. A divalent cation-independent protein serine/threonine phosphatase involved in the control of numerous cellular processes. Has a catalytic subunit of 36kDa, a subunit of 55kDa, and a regulatory subunit of 60kDa. Reported to associate with microtubules and to bind to purified tubulin polymerized by TAXOL™ Drug.Specifications
Concentration on Label | |
SDS-PAGE | |
PP2A1 | |
RUO | |
Native | |
≥90% |
In 50mM Tris-HCl, 14mM β-ME, 1mM benzamidine, 1mM EDTA, 100μM PMSF, 50% glycerol, pH 7.0. | |
5μg | |
Following initial thaw, aliquot and freeze (−20°C). | |
Bovine | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction