Learn More
EMD Millipore™ PP2A1, Bovine Kidney, Protein
Manufacturer:  Millipore Sigma 5395081UG
 View more versions of this product
Catalog No. 53-950-81UG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Concentration on Label
In 50mM Tris-HCl, 14mM β-ME, 1mM benzamidine, 1mM EDTA, 100μM PMSF, 50% glycerol, pH 7.0.
≥90%
PP2A1
RUO
No
Liquid
SDS-PAGE
1μg
Following initial thaw, aliquot and freeze (−20°C).
Bovine Kidney