Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Prion Protein, HisTag™, Human, Recombinant Protein, E. coli

Catalog No. 530006100UG Shop All MilliporeSigma Products
Click to view available options
Quantity:
100 μg

Catalog No. 53-000-6100UG

Supplier: MilliporeSigma™ 530006100UG

Only null left
Add to Cart
Add to Cart
Recombinant, human prion protein (amino acids 23-231) fused to a His·Tag™ sequence and expressed in E. coli. Useful as an antigen standard in immunochemical detection of Creutzfeldt-Jakob Disease (CJD).
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 10mM sodium acetate, pH 4.0.
Purification Method SDS-PAGE
Quantity 100 μg
Source E. coli
Immunogen Prion Protein
Storage Requirements Following initial thaw, aliquot and freeze (−20°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.