Learn More
EMD Millipore™ Protein G, Recombinant Protein, E. coli
Manufacturer:  Millipore Sigma 5393035MG
 View more versions of this product
Catalog No. 53-930-35MG
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Lyophilized Solid
SDS-PAGE
5mg
Protein G
RUO
H2O (5mg/mL) or Aqueous Buffers (5mg/mL).
≥98%
E. coli
Following reconstitution, aliquot and freeze (−20°C). Avoid freeze/thaw cycles. Stock solutions are stable for up to 6 months at −20°C.
Yes