Learn More
EMD Millipore™ pSTBlue-1 AccepTor Linearized Vectors
Manufacturer:  Millipore Sigma 705963
 View more versions of this product
Catalog No. 70-596-3
  Check Availability
Add to cart
Disclaimers:
For Research Use Only
Specifications
Description
The pSTBlue-1 is a general purpose vector with dual opposed T7 and SP6 promoters, both with amp and kan resistance, and an array of flanking restriction sites.