Learn More
MilliporeSigma™ pT7Blue Perfectly Blunt Cloning Kit
$430.00 - $683.00
Specifications
Vector pT7Blue Perfectly Blunt Vector
Includes pT7Blue Perfectly Blunt Vector; Positive Control Insert; End Conversion Mix; T4 DNA Ligase (Weiss units); Nuclease-free Water; NovaBlue Singles™ Competent Cells; SOC Medium; Test Plasmid for Transformation
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-189-4
View Documents
MilliporeSigma™
701894
40 rxn
Each for $683.00
Add to cart
70-189-3
View Documents
MilliporeSigma™
701893
20 rxn
Each for $430.00
Add to cart
Description
Description
The pT7Blue-3 vector is multi-purpose cloning vector featuring a versatile multiple cloning region, blue/white screening, a T7 promoter and dual kanamycin/ampicillin resistance. Restriction sites producing 4-base 3' overhangs are conveniently positioned at each end of the polylinker to facilitate the generation of unidirectional deletions using exonuclease III.
Specifications
Specifications
pT7Blue Perfectly Blunt Vector
pT7Blue Perfectly Blunt Vector; Positive Control Insert; End Conversion Mix; T4 DNA Ligase (Weiss units); Nuclease-free Water; NovaBlue Singles™ Competent Cells; SOC Medium; Test Plasmid for Transformation