Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma PTEN, HisTag and Stag Human Recombinant Protein, S. frugiperda
Manufacturer: MilliporeSigma 48140910UG
Description
Full-length, recombinant, human PTEN (GenBank target symbol = PTEN, ACC No. BC_005821) fused at N-terminus to a His·Tag and S·Tag sequences and expressed in S. frugiperda insect cells. PTEN is a tumor suppressor that is frequently found mutated in human cancers. It serves as a lipid phosphatase and negative regulator of the PI-3K/Akt pathway. Absence of functional PTEN results in constitutive activation of Akt and mTOR kinases.Specifications
Concentration on Label | |
In 150mM NaCl, 50mM Tris-HCl, 5mM DTT, 1mM EDTA, 50% glycerol, pH 7.5. | |
≥80% | |
S. frugiperda | |
Following initial thaw, aliquot and freeze (−70°C). | |
His·Tag™ and S·Tag™ | |
Yes |
Liquid | |
SDS-PAGE | |
10μg | |
PTEN | |
RUO | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title