Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ PureProteome™ Magnetic Beads
For rapid and reproducible isolation of proteins
$190.00 - $972.00
Specifications
Content And Storage | 2° to 8°C |
---|---|
For Use With (Application) | Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions |
Matrix | Magnetic Beads |
Catalog Number | Mfr. No. | Product Type | Quantity | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Particle Size | Price | Quantity & Availability | |||
MILSKMAGN01
|
MilliporeSigma™
LSKMAGN01 |
NHS flexiBind Magnetic bead system | 1 x 0.5mL | 10μm |
Each for $350.00
|
|
|||
MILSKMAGN04
|
MilliporeSigma™
LSKMAGN04 |
NHS flexiBind Magnetic bead system | 4 x 0.5mL | 10μm |
Each for $972.00
|
|
|||
LSKMAG1C02
|
MilliporeSigma™
LSKMAG1CBX02 |
Carboxy flexiBind Magnetic bead system | 2 x 1mL | 1μm |
Each for $190.00
|
|
|||
Description
Custom affinity bead generation for binding to target ligand
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Specifications
2° to 8°C | |
Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions | |
Magnetic Beads |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title