Learn More
MilliporeSigma™ Purospher™ STAR RP-18 Endcapped Hibar™ HR UHPLC Columns, 3 micron
For ultra-fast applications, where resolution, sensitivity and sample throughput are crucial
Manufacturer:  MilliporeSigma™ 1.50655.0001
 View more versions of this product
Catalog No. M1506550001
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Recommended for difficult samples where clogging and back-pressure present an issue
  • In UHPLC environment, speed is increased by a factor of 10
  • Solvent consumption is drastically cut (up to 85%)
  • Excellent peak shape for all types of acidic, basic and metal chelating analytes due to high purity silica
  • Extraordinary pH stability from pH 1.5 to 10.5 for an extremely wide application range
  • Enhanced sensitivity due to improved signal-to-noise ratio
Specifications
Specifications
Analytical Column
250 mm
3 μm
120 Å
330 m²/g
C18
17%
Yes
HR UHPLC Column
2.1 mm
8700 psi (600 bar)
1.5 to 10.5
1/Pk.
L1
Silica
Reversed Phase
Reversed Phase