Learn More
MilliporeSigma™ Qualitative Filter Papers
Supplier: MilliporeSigma™ FP10309000
Description
These 100% alpha-grade cotton cellulose fiber papers are ideal for clarifying and removing particles of 2.5μm and larger.
They're suitable for gravity and low suction filtration and can also be used in vacuum filtration systems. Available in 4.7 and 9.0cm diameters
Order Info
Come in boxes of 100.
Compliance
All papers meet ASTM E 832 for laboratory filter papers.
Specifications
6 to 8 μm | |
Suitable for separating precipitates: metal hydroxides. | |
Medium | |
9 cm |
240 sec. | |
103 | |
100 per box |