Learn More
MilliporeSigma™ Quant™ Test Strips Available on GSA/VA Contract for Federal Government customers only.
Easy-to-use test strips for field testing, preliminary semi-quantitative analysis or quality control
$115.50
Specifications
For Use With (Application) Field testing, preliminary semi-quantitative analysis or quality control
View More Specs
Includes: Any necessary reagents in dispenser bottles
Products 1
Catalog Number Mfr. No. Type Range Price Quantity & Availability  
Catalog Number Mfr. No. Type Range Price Quantity & Availability  
M1100440001 Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
1.10044.0001
Cyanide Test 1 to 30 ppm
Each for $115.50
Add to cart
Description
Description
Incorporate classical organic and inorganic reaction mechanisms that allow rapid, highly selective ion screening.
  • Ready to use directly or comes as a kit; all necessary detection chemicals, buffers, masking agents and stabilizers are incorporated into the reaction pads or in the kits
  • For most applications, pretreatment is unnecessary when other ionic species are present at concentrations less than 1000ppm
  • Use appropriate dilution procedures to test solutions with concentrations above the upper range of the test strip
  • Test strips are supplied in sturdy aluminum cylinders labeled with instructions, color comparison scale and expiration date
Order Info
For additional analytes not featured here, contact your Fisher Scientific Customer Service Representative for ordering information.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Field testing, preliminary semi-quantitative analysis or quality control