Learn More
MilliporeSigma™ Quantitative Filter Papers: Ashless Circles
Manufacturer:  MilliporeSigma™ FP22109000
 View more versions of this product
Catalog No. FP22109000
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Specifications
Specifications
Circle
2.5μm
Provides maximum degree of particle retention. Filters finely grained precipitates.
Fine
9 cm
Cellulose Fiber
DIN 53137, 40 seconds; Herzberg, 1500 seconds
221
100/Pk.