Learn More
MilliporeSigma™ Quantitative Filter Papers: Ashless Circles
Manufacturer:  MilliporeSigma™ FP22209000
 View more versions of this product
Catalog No. FP22209000
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
1Filtration speed conditions. DIN 53137, time for 10mL to pass through filter; Herzberg, time for 100mL to pass through 10cm2 filter paper with 10cm constant head pressure of water.
Specifications
Specifications
Circle
3μm
Fastest for fine particle retention. Similar to Grade 221, but with twice the flowrate. Use for filtering extremely difficult and finely grained precipitates.
Fine
9 cm
Cellulose Fiber
DIN 53137, 75 seconds; Herzberg, 750 seconds
222
100/Pk.