Learn More
MilliporeSigma™ Quantitative Filter Papers: Ashless Circles
Manufacturer:  MilliporeSigma™ FP22304700
 View more versions of this product
Catalog No. FP22304700
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Circle
8μm
Gravimetric analysis, filtration of solutions prior to atomic absorption analysis, air pollution monitoring and gas detection.
Medium
4.7 cm
Cellulose Fiber
DIN 53137, 48 seconds; Herzberg, 480 seconds
223
100/Pk.