Learn More
MilliporeSigma™ Quantitative Filter Papers: Ashless Circles
Manufacturer:  MilliporeSigma™ FP22309000
 View more versions of this product
Catalog No. FP22309000
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
1Filtration speed conditions. DIN 53137, time for 10mL to pass through filter; Herzberg, time for 100mL to pass through 10cm2 filter paper with 10cm constant head pressure of water.
Specifications
Specifications
Circle
8μm
Gravimetric analysis, filtration of solutions prior to atomic absorption analysis, air pollution monitoring and gas detection.
Medium
9 cm
Cellulose Fiber
DIN 53137, 48 seconds; Herzberg, 480 seconds
223
100/Pk.