Learn More
MilliporeSigma™ Quantitative Filter Papers: Ashless Circles
Manufacturer:  MilliporeSigma™ FP22404700
 View more versions of this product
Catalog No. FP22404700
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Circle
16μm
Precipitates collection, soil analysis, inorganic analysis and air pollution particle collection.
Medium
4.7 cm
Cellulose Fiber
DIN 53137, 13 seconds; Herzberg, 140 seconds
224
100/Pk.