Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Quantitative Filter Papers: Ashless Circles

Specifications

Quantity 100/Pk.
Format Circle
Material Cellulose Fiber
View More Specs

Products 9
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
FP22309000
View Documents
MilliporeSigma™
FP22309000
Grade: 223; 8μm particle retention; Dia.: 90mm Grade: 223; 8μm particle retention; Dia.: 90mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP22209000
View Documents
MilliporeSigma™
FP22209000
Grade: 222; 3μm particle retention; Dia.: 90mm Grade: 222; 3μm particle retention; Dia.: 90mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP22104700
View Documents
MilliporeSigma™
FP22104700
Grade: 221; 2.5μm particle retention; Dia.: 47mm Grade: 221; 2.5μm particle retention; Dia.: 47mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FP22204700
View Documents
MilliporeSigma™
FP22204700
Grade: 222; 3μm particle retention; Dia.: 47mm Grade: 222; 3μm particle retention; Dia.: 47mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FP22109000
View Documents
MilliporeSigma™
FP22109000
Grade: 221; 2.5μm particle retention; Dia.: 90mm Grade: 221; 2.5μm particle retention; Dia.: 90mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP22304700
View Documents
MilliporeSigma™
FP22304700
Grade: 223; 8μm particle retention; Dia.: 47mm Grade: 223; 8μm particle retention; Dia.: 47mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FP22404700
View Documents
MilliporeSigma™
FP22404700
Grade: 224; 16μm particle retention; Dia.: 47mm Grade: 224; 16μm particle retention; Dia.: 47mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
FP22409000
View Documents
MilliporeSigma™
FP22409000
Grade: 224; 16μm particle retention; Dia.: 90mm Grade: 224; 16μm particle retention; Dia.: 90mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP22504700
View Documents
MilliporeSigma™
FP22504700
Grade: 225; 20 to 25μm particle retention; Dia.: 47mm Grade: 225; 20 to 25μm particle retention; Dia.: 47mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

1Filtration speed conditions. DIN 53137, time for 10mL to pass through filter; Herzberg, time for 100mL to pass through 10cm2 filter paper with 10cm constant head pressure of water.

Specifications

Specifications

100/Pk.
Circle
Cellulose Fiber
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit