Learn More
MilliporeSigma™ Quantitative Filter Papers: Hardened Ashless Circles
Supplier:  MilliporeSigma™ FP23509000
 View more versions of this product
Catalog No. FP23509000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View alternate product
Description
Description
Specifications
Specifications
Disc
20 to 25 μm
Protein determinations. Analyzing fiber in animal foodstuffs, gelatin in milk and cream. For Si determination in steel and pig iron.
Coarse
Cellulose Fiber
DIN 53137, 9 seconds; Herzberg, 40 seconds
235
9 cm
Product Suggestions
Product Suggestions