Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Quantitative Filter Papers: Hardened Ashless Circles

Specifications

Format Circle
Material Cellulose Fiber
View More Specs

Products 4
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
FP23309000
View Documents
MilliporeSigma™
FP23309000
Grade: 233; Diameter: 9.0cm Grade: 233; Diameter: 9.0cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP23509000
View Documents
MilliporeSigma™
FP23509000
Grade: 235; Diameter: 9.0cm Grade: 235; Diameter: 9.0cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP23504700
View Documents
MilliporeSigma™
FP23504700
Grade: 235; Diameter: 4.7cm Grade: 235; Diameter: 4.7cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
FP23304700
View Documents
MilliporeSigma™
FP23304700
Grade: 233; Diameter: 4.7cm Grade: 233; Diameter: 4.7cm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

1Filtration speed conditions. DIN 53137, time for 10mL to pass through filter; Herzberg, time for 100mL to pass through 10cm2 filter paper with 10cm constant head pressure of water.

Specifications

Specifications

Circle
Cellulose Fiber
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit