EMD Millipore ReBlot Plus Kit and Reagents Available on GSA/VA Contract for Federal Government customers only.

Blots can be stripped of antibodies in approximately 15 minutes at room temperature

Manufacturer: EMD Millipore 2502

 View more versions of this product

Catalog No. 2502MI

  Check Availability
Add to Cart

    Specifications

    Kit Contents 25mL Mild Antibody stripping solution 10X
    Product Type ReBlot Plus 10X mild Antibody stripping solution
    Applications Western blotting
    Quantity 50mL
    Storage Requirements 2° to 8°C
    Concentration 2X
    Certifications/Compliance 29 CFR 1910.1200, 67 548 EEC, 1999 45 EC

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit