Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Reflectoquant™ Iron Test Strips

To determine the presence of residues of iron

$113.40

Specifications

Type Test Strip
For Use With (Application) Ideal for measuring iron(II) ions in surface water, seawater, industrial water, wastewater and mineral water
For Use With (Equipment) RQflex™ 10 and RQflex™ Plus 10 reflectometers
No. of Tests 50
Storage Requirements 15°C to 25°C
View More Specs

Includes: Tube containing 50 test strips, 1 bar-code strip

Products 1
Catalog Number Mfr. No. Range Graduations Test Method Price Quantity & Availability  
Catalog Number Mfr. No. Range Graduations Test Method Price Quantity & Availability  
M1169820001
View Documents
MilliporeSigma™
1.16982.0001
0.5 to 20mg/L 0.5 to 20mg/L Triazine
Each for $113.40
Add to cart
 
Description

Description

  • Triazine reaction method
  • Barcode calibration for reliable, quantitative results
  • Low analysis costs

Reclose the tube containing the test strips immediately after use.

For food and water analysis

Specifications

Specifications

Test Strip
RQflex™ 10 and RQflex™ Plus 10 reflectometers
15°C to 25°C
Ideal for measuring iron(II) ions in surface water, seawater, industrial water, wastewater and mineral water
50
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit