EMD Millipore Replacement Parts for Stirred Cells Available on GSA/VA Contract for Federal Government customers only.
Suitable for use with Amicon™ stirred cell apparatus
Manufacturer: EMD Millipore 8400SCPKIT
 View more versions of this product
Catalog No. 8400SCPKIT
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Amicon™ 8400 series stirred cell apparatus
    Includes Cap assembly (1524901), stirrer assembly (15247) and base (210000)
    Type Replacement Part
  • Description & Specifications
Related Products
Product Content Feedback