EMD Millipore Replacement Parts for Stirred Cells Available on GSA/VA Contract for Federal Government customers only.
Suitable for use with Amicon™ stirred cell apparatus
Manufacturer: EMD Millipore 8000SCPKIT
 View more versions of this product
Catalog No. 8000SCPKIT
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Amicon™ 8000 series stirred cell apparatus
    Includes 2 O-rings (2122S, 2118S, 2015S), tubing (4500202), tubing fitting assembly (15025AM) and membrane holder (2214801)
    Type Replacement Part
  • Description & Specifications
Related Products
Product Content Feedback